Learn More
MSC KleenGuard Disposable Coveralls, Liquid and Particle Protection, X-Large, 25 Pack
Catalog No. 5000686108
Encompass
KleenGuard™ A35 Disposable Coveralls provide comprehensive protection against hazardous liquid splashes and dry particles. Constructed from white, microporous film-laminated material, these coveralls are designed for various industrial and manufacturing applications.
  • Features a hood, boots, zip front, and elastic wrists
  • Provides liquid and particle protection
  • Meets ANSI/ISEA 101-1996 Standards
  • Ideal for hazardous liquid handling, pressure washing, maintenance, and general manufacturing
Catalog No. 50-006-86108 Supplier MSC Supplier No. 54549118
KleenGuard™ A35 Disposable Coveralls provide comprehensive protection against hazardous liquid splashes and dry particles. Constructed from white, microporous film-laminated material, these coveralls are designed for various industrial and manufacturing applications.
  • Features a hood, boots, zip front, and elastic wrists
  • Provides liquid and particle protection
  • Meets ANSI/ISEA 101-1996 Standards
  • Ideal for hazardous liquid handling, pressure washing, maintenance, and general manufacturing