Learn More
MSC KleenGuard Lab Coat, X-Large, Microporous Film Laminate, White, 30 Pack
Catalog No. 5000796508
Encompass
This lab coat is constructed from microporous film laminate, offering protection and comfort for various non-cleanroom applications.
  • Features an open cuff style
  • Secures with snap closures
  • Designed for non-cleanroom ISO class environments
  • Equipped with two pockets for convenience
Catalog No. 50-007-96508 Supplier MSC Supplier No. 15210222
This lab coat is constructed from microporous film laminate, offering protection and comfort for various non-cleanroom applications.
  • Features an open cuff style
  • Secures with snap closures
  • Designed for non-cleanroom ISO class environments
  • Equipped with two pockets for convenience