Learn More
MSC KleenGuard Lab Coat, X-Large, Microporous Film, White, 30/Pk.
Catalog No. 5000765608
Encompass
KleenGuard A40 Liquid and Particle Protection Lab Coats are knee-length with surged seams, elastic wrists, a five-snap closure, and no pockets. They are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
  • Surged seams, elastic wrists, five-snap closure, and no pockets
  • Made of breathable, microporous film
  • Provides a better liquid barrier compared to DuPont Tyvek
  • Passes NFPA 99 criteria for antistatic materials
  • Passes ASTM F1670 testing for penetration of blood and bodily fluids
  • Meets ANSI/ISEA 101-1996 sizing standards
Catalog No. 50-007-65608 Supplier MSC Supplier No. 70699053
KleenGuard A40 Liquid and Particle Protection Lab Coats are knee-length with surged seams, elastic wrists, a five-snap closure, and no pockets. They are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
  • Surged seams, elastic wrists, five-snap closure, and no pockets
  • Made of breathable, microporous film
  • Provides a better liquid barrier compared to DuPont Tyvek
  • Passes NFPA 99 criteria for antistatic materials
  • Passes ASTM F1670 testing for penetration of blood and bodily fluids
  • Meets ANSI/ISEA 101-1996 sizing standards