Learn More
MSC KleenGuard OTG Eyewear, Black Frame, Smoke Mirror Anti-Fog, 1 ea
Catalog No. 5000792527
Encompass
Stylish safety eyewear designed to fit over most prescription eyeglasses. They are suitable for individuals who wear prescription glasses and require safety eye protection.
  • Fits over most glasses
  • Integral side shields for side impact protection without obstructing peripheral vision
  • Comfortable fit
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
  • Standard anti-fog/scratch-resistant coating
  • Meets ANSI Z87.1+ and CSA Z94.3 standards for high impact
  • Deep integrated brow design for secure fit and reduced gaps
  • Strong and durable construction
Catalog No. 50-007-92527 Supplier MSC Supplier No. 57685083
Stylish safety eyewear designed to fit over most prescription eyeglasses. They are suitable for individuals who wear prescription glasses and require safety eye protection.
  • Fits over most glasses
  • Integral side shields for side impact protection without obstructing peripheral vision
  • Comfortable fit
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
  • Standard anti-fog/scratch-resistant coating
  • Meets ANSI Z87.1+ and CSA Z94.3 standards for high impact
  • Deep integrated brow design for secure fit and reduced gaps
  • Strong and durable construction