Learn More
MSC KleenGuard Safety Glasses, Anti-Fog, Polycarbonate, Half-Framed, ANSI Z87.1, 1 ea
Catalog No. 5000787663
Encompass
These safety glasses feature a modern and sleek lightweight design, equipped with anti-fog and scratch-resistant polycarbonate lenses for reliable eye protection. They offer 99.9% UVA/UVB/UVC protection and meet ANSI Z87.1 standards, making them suitable for various industrial and laboratory applications.
  • Anti-fog and scratch-resistant lenses
  • Half-framed black frame with wrap-around design
  • Clear polycarbonate lenses
  • Non-adjustable temples
  • Provides 99.9% UVA/UVB/UVC protection
  • Meets ANSI Z87.1 standards
  • Cord Connect allows for easy attachment of earplug cords
Catalog No. 50-007-87663 Supplier MSC Supplier No. 02760338
These safety glasses feature a modern and sleek lightweight design, equipped with anti-fog and scratch-resistant polycarbonate lenses for reliable eye protection. They offer 99.9% UVA/UVB/UVC protection and meet ANSI Z87.1 standards, making them suitable for various industrial and laboratory applications.
  • Anti-fog and scratch-resistant lenses
  • Half-framed black frame with wrap-around design
  • Clear polycarbonate lenses
  • Non-adjustable temples
  • Provides 99.9% UVA/UVB/UVC protection
  • Meets ANSI Z87.1 standards
  • Cord Connect allows for easy attachment of earplug cords