Learn More
MSC KleenGuard Safety Glasses, Anti-Fog and Scratch-Resistant, Polycarbonate, 1 ea
Catalog No. 5000768143
Encompass
This product is a pair of full-framed safety glasses with clear polycarbonate lenses. They feature anti-fog and scratch-resistant coatings, a silver frame with adjustable temples, and are part of the Visio Series. These safety glasses meet ANSI Z87.1+ standards.
  • Clear lens
  • Anti-fog coating improves visibility under hot and humid conditions
  • Scratch-resistant
  • Full-framed design
  • Wrap around frame design
  • Silver frame color
  • Universal size
  • Visio series
  • Polycarbonate lens material
  • Adjustable temples
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Three-point extendable temples
  • Plastic frame material
Catalog No. 50-007-68143 Supplier MSC Supplier No. 05731419
This product is a pair of full-framed safety glasses with clear polycarbonate lenses. They feature anti-fog and scratch-resistant coatings, a silver frame with adjustable temples, and are part of the Visio Series. These safety glasses meet ANSI Z87.1+ standards.
  • Clear lens
  • Anti-fog coating improves visibility under hot and humid conditions
  • Scratch-resistant
  • Full-framed design
  • Wrap around frame design
  • Silver frame color
  • Universal size
  • Visio series
  • Polycarbonate lens material
  • Adjustable temples
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Three-point extendable temples
  • Plastic frame material