Learn More
MSC KleenGuard Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, ANSI Z87.1, 1 ea
Catalog No. 5000787839
Encompass
These safety glasses are designed with anti-fog and scratch-resistant clear polycarbonate lenses, featuring a full-framed, wrap-around design. They are equipped with a black frame and non-adjustable black temples, meeting ANSI Z87.1+ standards and offering UV protection.
  • Anti-fog and scratch-resistant lenses
  • Polycarbonate lens material
  • Clear lenses for optimal visibility
  • Full-framed, wrap-around design
  • Black frame and non-adjustable temples
  • ANSI Z87.1+ compliant
  • Provides UV protection
  • Universal size for broad compatibility
Catalog No. 50-007-87839 Supplier MSC Supplier No. 43422898
These safety glasses are designed with anti-fog and scratch-resistant clear polycarbonate lenses, featuring a full-framed, wrap-around design. They are equipped with a black frame and non-adjustable black temples, meeting ANSI Z87.1+ standards and offering UV protection.
  • Anti-fog and scratch-resistant lenses
  • Polycarbonate lens material
  • Clear lenses for optimal visibility
  • Full-framed, wrap-around design
  • Black frame and non-adjustable temples
  • ANSI Z87.1+ compliant
  • Provides UV protection
  • Universal size for broad compatibility