Learn More
MSC KleenGuard Safety Glasses, Polycarbonate, IRUV Shade 5.0, 1 Each
Catalog No. 5000768238
Encompass
These safety glasses feature an IRUV 5.0 polycarbonate green lens and a black frame, designed to provide eye protection during tasks such as medium cutting and brazing. They meet ANSI Z87.1+2015 standards for high impact protection.
  • Sporty, flexible, and lightweight design for comfort
  • Patented nosepiece with integrated ridges to divert sweat
  • Integrated neck cord to reduce damage risk
  • Hardcoated lenses for scratch resistance and extended life
  • Provides 99.9% UVA/UVB/UVC protection
  • IRUV Shade 5 lenses protect against infrared and ultraviolet radiation
Catalog No. 50-007-68238 Supplier MSC Supplier No. 00311308
These safety glasses feature an IRUV 5.0 polycarbonate green lens and a black frame, designed to provide eye protection during tasks such as medium cutting and brazing. They meet ANSI Z87.1+2015 standards for high impact protection.
  • Sporty, flexible, and lightweight design for comfort
  • Patented nosepiece with integrated ridges to divert sweat
  • Integrated neck cord to reduce damage risk
  • Hardcoated lenses for scratch resistance and extended life
  • Provides 99.9% UVA/UVB/UVC protection
  • IRUV Shade 5 lenses protect against infrared and ultraviolet radiation