Learn More
MSC KleenGuard Safety Glasses, Scratch-Resistant, Blue Mirror, Polycarbonate, 1 ea
Catalog No. 5000769667
Encompass
These V30 Nemesis safety glasses feature blue mirror polycarbonate lenses and a black full-framed design. They offer 99.9% UVA/UVB/UVC protection and meet ANSI Z87.1+ standards, ensuring reliable eye protection in various work environments.
  • Scratch-resistant polycarbonate lenses
  • Blue mirror lenses with a full-framed design
  • Provides 99.9% UVA/UVB/UVC protection
  • Meets ANSI Z87.1+ standards
  • Designed to be extremely flexible
  • Channels sweat away from the eyes and prevents slippage
  • Includes a neck cord
  • Eligible for the EOL recycling program
Catalog No. 50-007-69667 Supplier MSC Supplier No. 59829028
These V30 Nemesis safety glasses feature blue mirror polycarbonate lenses and a black full-framed design. They offer 99.9% UVA/UVB/UVC protection and meet ANSI Z87.1+ standards, ensuring reliable eye protection in various work environments.
  • Scratch-resistant polycarbonate lenses
  • Blue mirror lenses with a full-framed design
  • Provides 99.9% UVA/UVB/UVC protection
  • Meets ANSI Z87.1+ standards
  • Designed to be extremely flexible
  • Channels sweat away from the eyes and prevents slippage
  • Includes a neck cord
  • Eligible for the EOL recycling program