Learn More
MSC KleenGuard Safety Glasses, Smoke Polycarbonate Lens, Scratch-Resistant, 12 PK
Catalog No. 5000793535
Encompass
KleenGuard Purity Safety Eyewear offers stylish yet economical eye protection. Designed with padded temples for enhanced comfort, these safety glasses feature smoke lenses ideal for outdoor use, and are hardcoated for superior scratch resistance, ensuring prolonged life. They provide 99.9% UVA/UVB/UVC protection, helping to prevent eye damage.
  • Padded temples for enhanced comfort
  • Smoke lenses ideal for outdoor use
  • Hardcoated for scratch resistance
  • Polycarbonate lenses
  • Provides 99.9% UVA/UVB/UVC protection
  • Complies with ANSI/ISEA Z87.1+ standard
  • Certified to meet CSA Z94.3 requirements
Catalog No. 50-007-93535 Supplier MSC Supplier No. 57685356
KleenGuard Purity Safety Eyewear offers stylish yet economical eye protection. Designed with padded temples for enhanced comfort, these safety glasses feature smoke lenses ideal for outdoor use, and are hardcoated for superior scratch resistance, ensuring prolonged life. They provide 99.9% UVA/UVB/UVC protection, helping to prevent eye damage.
  • Padded temples for enhanced comfort
  • Smoke lenses ideal for outdoor use
  • Hardcoated for scratch resistance
  • Polycarbonate lenses
  • Provides 99.9% UVA/UVB/UVC protection
  • Complies with ANSI/ISEA Z87.1+ standard
  • Certified to meet CSA Z94.3 requirements