Learn More
MSC KleenGuard Safety Goggles, Chemical Splash, Anti-Fog, Clear Polycarbonate, 1 ea
Catalog No. 5000677833
Encompass
These safety goggles feature clear polycarbonate lenses, providing protection against chemical splash and dust. They are equipped with anti-fog and scratch-resistant coatings to maintain visibility, and an indirect vent system for comfortable wear.
  • Clear lens type
  • Anti-fog and scratch-resistant lens coating
  • Indirect vent system
  • Protects against dust and splash
  • Polycarbonate lens material
  • Clear polycarbonate frame
  • Universal size, fits over prescription glasses
  • Offers UV protection
  • Meets ANSI Z87.1+ standards
  • Anti-fog coatings improve visibility under hot and humid conditions
Catalog No. 50-006-77833 Supplier MSC Supplier No. 08270514
These safety goggles feature clear polycarbonate lenses, providing protection against chemical splash and dust. They are equipped with anti-fog and scratch-resistant coatings to maintain visibility, and an indirect vent system for comfortable wear.
  • Clear lens type
  • Anti-fog and scratch-resistant lens coating
  • Indirect vent system
  • Protects against dust and splash
  • Polycarbonate lens material
  • Clear polycarbonate frame
  • Universal size, fits over prescription glasses
  • Offers UV protection
  • Meets ANSI Z87.1+ standards
  • Anti-fog coatings improve visibility under hot and humid conditions