Learn More
MSC KleenGuard Sleeve Protector, Polyethylene, Chemical Resistant, 21 in, 200/Case
Catalog No. 5000796017
Encompass
KleenGuard A70 Sleeve Protectors offer chemical spray protection for various industrial and laboratory applications. These disposable sleeves provide resistance against many water-based liquids, greases, oils, and liquid chemical splashes, ensuring reliable arm protection in demanding environments.
  • Chemical spray protection
  • Resists water-based liquids, greases, and oils
  • Constructed from polyethylene coated polypropylene
  • Overall length of 21 in
  • Thickness of 1.5 mil
  • Elastic openings at both ends
  • Passes NFPA 99 for antistatic materials
  • Disposable
  • Latex-free
Catalog No. 50-007-96017 Supplier MSC Supplier No. 08839367
KleenGuard A70 Sleeve Protectors offer chemical spray protection for various industrial and laboratory applications. These disposable sleeves provide resistance against many water-based liquids, greases, oils, and liquid chemical splashes, ensuring reliable arm protection in demanding environments.
  • Chemical spray protection
  • Resists water-based liquids, greases, and oils
  • Constructed from polyethylene coated polypropylene
  • Overall length of 21 in
  • Thickness of 1.5 mil
  • Elastic openings at both ends
  • Passes NFPA 99 for antistatic materials
  • Disposable
  • Latex-free