Learn More
MSC Rochester Midland Corporation Glass & Surface Cleaner, Liquid Concentrate, Mild Fragrance, pH 7.0
Catalog No. 502835975
Encompass
This concentrated cleaner is designed for glass and various surfaces, formulated to be biobased, ammonia-free, and non-streaking. With a mild fragrance and a neutral pH of 7.0, it effectively cleans and removes a wide range of residues, leaving surfaces clear and bright. It is suitable for use on glass, mirrors, porcelain, and monitors.
  • No ammonia or alcohol
  • Biodegradable formulation
  • Contains no hazardous ingredients
  • Non-corrosive and NPE-free
  • Mild fragrance for a pleasant cleaning experience
  • Neutral pH of 7.0 for versatile cleaning
  • Effectively removes dust, oil, grease, and streaks
  • Ideal for glass, mirrors, porcelain, and monitor screens
  • Concentrated for dilutable use
Catalog No. 50-283-5975 Supplier MSC Supplier No. 63333330
This concentrated cleaner is designed for glass and various surfaces, formulated to be biobased, ammonia-free, and non-streaking. With a mild fragrance and a neutral pH of 7.0, it effectively cleans and removes a wide range of residues, leaving surfaces clear and bright. It is suitable for use on glass, mirrors, porcelain, and monitors.
  • No ammonia or alcohol
  • Biodegradable formulation
  • Contains no hazardous ingredients
  • Non-corrosive and NPE-free
  • Mild fragrance for a pleasant cleaning experience
  • Neutral pH of 7.0 for versatile cleaning
  • Effectively removes dust, oil, grease, and streaks
  • Ideal for glass, mirrors, porcelain, and monitor screens
  • Concentrated for dilutable use