Learn More
MSC Klein Tools Safety Helmet Visor, Clear, Anti-Fog, 1 ea
Catalog No. 5000796651
Encompass
This safety helmet visor is constructed from lightweight polycarbonate with vents for adequate ventilation and long-term comfort. It features durable anti-fog, scratch-resistant, and UV-protective coatings, specifically designed for use with compatible safety helmets.
  • Optical Class-1 clear lens tested for minimal distortion
  • Half-face, wrap-around design maintains a low profile
  • Superior impact protection meets ANSI Z87.1+, CSA Z94.3-20, and MIL-PRF-32432A standards
  • Durable anti-fog coating resists wear, even with dust masks
  • Specially treated lenses offer enhanced scratch resistance
  • Blocks 99.9% of harmful UV-A, UV-B, and UV-C rays (ANSI Z87.1 U6 standard)
  • Visor locks in up and down positions for desired placement
Catalog No. 50-007-96651 Supplier MSC Supplier No. 03920402
This safety helmet visor is constructed from lightweight polycarbonate with vents for adequate ventilation and long-term comfort. It features durable anti-fog, scratch-resistant, and UV-protective coatings, specifically designed for use with compatible safety helmets.
  • Optical Class-1 clear lens tested for minimal distortion
  • Half-face, wrap-around design maintains a low profile
  • Superior impact protection meets ANSI Z87.1+, CSA Z94.3-20, and MIL-PRF-32432A standards
  • Durable anti-fog coating resists wear, even with dust masks
  • Specially treated lenses offer enhanced scratch resistance
  • Blocks 99.9% of harmful UV-A, UV-B, and UV-C rays (ANSI Z87.1 U6 standard)
  • Visor locks in up and down positions for desired placement