Learn More
Sklar Instruments Sklaren(TM) Powder Detergent, Sklar , Powder Chemical (Sodium Dodecyl Benzene Sulfonate, Sodium Bicarbonate, Sodium Tripolyphosphate). 5 lb. Pail Sklaren
SDP
Catalog No. NC9879102 Shop All Sklar Instruments Products
Encompass_Preferred
Sklar Kleen(TM) Powder Detergent, Sklar, Powder Chemical (Sodium Dodecyl Benzene Sulfonate, Sodium Bicarbonate, Sodium Tripolyphosphate). 5 lb. Pail Sklar Kleen Powder Detergent SDS Concentrated For Manual and Ultrasonic Cleaning. EA.
Catalog No. NC9879102 Supplier Sklar Instruments Supplier No. 101607
Sklar Kleen(TM) Powder Detergent, Sklar, Powder Chemical (Sodium Dodecyl Benzene Sulfonate, Sodium Bicarbonate, Sodium Tripolyphosphate). 5 lb. Pail Sklar Kleen Powder Detergent SDS Concentrated For Manual and Ultrasonic Cleaning. EA.