Learn More
Qiagen Beverly Inc KLENOW (3 - 5 EXO-)
Catalog No. 50305912
Encompass
DNA Polymerase; Klenow (3'-5' Exo-) High Conc.; Conc. 50,000 U/ml; 10,000 U; incl. 10X buffer
Catalog No. 50-305-912 Supplier Qiagen Beverly Inc Supplier No. P7010HCL
DNA Polymerase; Klenow (3'-5' Exo-) High Conc.; Conc. 50,000 U/ml; 10,000 U; incl. 10X buffer