Learn More
New England Biolabs, Inc. Klenow Fragment (3'-5' exo-) – 1000 units
SDP
Supplier:  New England Biolabs, Inc. M0212L
Encompass
Klenow Fragment (3' to 5' exo-) is an N-terminal truncation of DNA Polymerase I which retains polymerase activity, but has lost the 5' to 3' exonuclease activity and has mutations (D355A, E357A) which abolish the 3' to 5' exonuclease activity.
Catalog No. 50-811-626