Learn More
New England Biolabs, Inc. Klenow Fragment (3'-5' exo-) – 200 units
SDP
Catalog No. 50811628
Encompass
Klenow Fragment (3' to 5' exo-) is an N-terminal truncation of DNA Polymerase I which retains polymerase activity, but has lost the 5' to 3' exonuclease activity and has mutations (D355A, E357A) which abolish the 3' to 5' exonuclease activity.
Catalog No. 50-811-628 Supplier New England Biolabs, Inc. Supplier No. M0212S
Klenow Fragment (3' to 5' exo-) is an N-terminal truncation of DNA Polymerase I which retains polymerase activity, but has lost the 5' to 3' exonuclease activity and has mutations (D355A, E357A) which abolish the 3' to 5' exonuclease activity.