Learn More
Ab Bioscience LLC KlenTaq1 DNA Polymerase, 100ul
Catalog No. NC9707348
Encompass
KlenTaq1-Increments of 100 µl enzyme (sufficient for 2000 x 25 µl reactions up to 1 kb) KlenTaq1 is a 5'-exonuclease deficient Taq polymerase (an N-terminal deletion of Taq) leaving highly active and even more heat-stable DNA polymerase activity.
Catalog No. NC9707348 Supplier Ab Bioscience LLC Supplier No. 1001
KlenTaq1-Increments of 100 µl enzyme (sufficient for 2000 x 25 µl reactions up to 1 kb) KlenTaq1 is a 5'-exonuclease deficient Taq polymerase (an N-terminal deletion of Taq) leaving highly active and even more heat-stable DNA polymerase activity.