Learn More
Ecolab Klercide 70/30 Pharma Disinfectant, Sterile, 70% Ethanol with IPA, 6 Units Per Case
Catalog No. NC3346023
Encompass
Klercide 70/30 Pharma Ethanol is a highly effective sterile disinfectant specifically designed for critical cleanroom environments. Composed of 70% ethanol and isopropyl alcohol (IPA), it ensures minimal residue and high purity. Its patented SteriShield™ Delivery System maintains sterility, preventing contamination. With broad-spectrum efficacy against bacteria, fungi, and viruses, it is manufactured under cGMP standards, making it ideal for aseptic processing.
Catalog No. NC3346023 Supplier Ecolab Supplier No. 6000198
Klercide 70/30 Pharma Ethanol is a highly effective sterile disinfectant specifically designed for critical cleanroom environments. Composed of 70% ethanol and isopropyl alcohol (IPA), it ensures minimal residue and high purity. Its patented SteriShield™ Delivery System maintains sterility, preventing contamination. With broad-spectrum efficacy against bacteria, fungi, and viruses, it is manufactured under cGMP standards, making it ideal for aseptic processing.