Learn More
Ecolab Klercide Sporicidal Active Chlorine, Sterile Sodium Hypochlorite Solution, 6 Bottles x 32 oz
Catalog No. NC2209437
Encompass
The Ecolab Klercide Sporicidal Active Chlorine is a sterile, ready-to-use sodium hypochlorite solution designed for critical cleanroom environments. It effectively combats a range of microbial contaminants, including bacterial spores and viruses, without additional activators. Its unique SteriShield Delivery System minimizes operator risks, making it superior to standard disinfectants. The product ensures high purity and sterility, meeting stringent cleanroom standards.
This item is not returnable. View return policy
Catalog No. NC2209437 Supplier Ecolab Supplier No. 6600110
The Ecolab Klercide Sporicidal Active Chlorine is a sterile, ready-to-use sodium hypochlorite solution designed for critical cleanroom environments. It effectively combats a range of microbial contaminants, including bacterial spores and viruses, without additional activators. Its unique SteriShield Delivery System minimizes operator risks, making it superior to standard disinfectants. The product ensures high purity and sterility, meeting stringent cleanroom standards.