Learn More
Ecolab KLERCIDE SPORICIDAL ACTIVE CHLORINE - 4-1GL (2 bag)
Catalog No. NC2948964
Encompass
Versatile sporicide with a broad spectrum of activity and specifically formulated to help control microbial contamination most likely to be found in a cleanroom environment, including difficult to kill bacterial spores. Klercide Sporicidal Active Chlorine* is a sterile sodium hypochlorite solution containing 0.55% (5500 ppm) available chlorine as a ready-to-use solution.The product is 0.2 micron filtered, double bagged and filled in an ISO Class 7 cleanroom.
Catalog No. NC2948964 Supplier Ecolab Supplier No. 6600108
Versatile sporicide with a broad spectrum of activity and specifically formulated to help control microbial contamination most likely to be found in a cleanroom environment, including difficult to kill bacterial spores. Klercide Sporicidal Active Chlorine* is a sterile sodium hypochlorite solution containing 0.55% (5500 ppm) available chlorine as a ready-to-use solution.The product is 0.2 micron filtered, double bagged and filled in an ISO Class 7 cleanroom.