Learn More
Ecolab Ecolab Klercide 70/30 IPA Sterile, 70% IPA, Endotoxin Tested, 16 oz
Catalog No. NC2380658
Encompass
Ecolab Klercide 70/30 IPA Sterile is a high-grade disinfectant designed for critical cleanroom environments. Its unique formulation includes 70% Isopropyl Alcohol blended with Water for Injection, providing broad-spectrum bactericidal, fungicidal, and virucidal efficacy. Differentiated by the SteriShield™ Delivery System, it ensures sterility for up to three months post-opening. Ideal for aseptic processing areas and suitable for multiple cleanroom grades, it enhances sanitation practices effectively.
Catalog No. NC2380658 Supplier Ecolab Supplier No. 6600137
Ecolab Klercide 70/30 IPA Sterile is a high-grade disinfectant designed for critical cleanroom environments. Its unique formulation includes 70% Isopropyl Alcohol blended with Water for Injection, providing broad-spectrum bactericidal, fungicidal, and virucidal efficacy. Differentiated by the SteriShield™ Delivery System, it ensures sterility for up to three months post-opening. Ideal for aseptic processing areas and suitable for multiple cleanroom grades, it enhances sanitation practices effectively.