Learn More
Ecolab Klercide Cleanroom Quat, Broad-Spectrum Efficacy, Minimal Residue Formulation, 6 x 32 oz
Catalog No. NC2948778
Encompass
Klercide Cleanroom Quat is a sterile quaternary ammonium compound solution designed for sanitization in pharmaceutical cleanrooms. Its low-residue formulation minimizes cleaning requirements, enhancing labor efficiency and production time. Effective against a broad spectrum of microorganisms including bacteria and viruses, it is supplied with a SteriShield Delivery System to maintain sterility. Ideal for hard, non-porous surfaces in controlled environments, Klercide ensures a clean and safe workspace.
Catalog No. NC2948778 Supplier Ecolab Supplier No. 6600149
Klercide Cleanroom Quat is a sterile quaternary ammonium compound solution designed for sanitization in pharmaceutical cleanrooms. Its low-residue formulation minimizes cleaning requirements, enhancing labor efficiency and production time. Effective against a broad spectrum of microorganisms including bacteria and viruses, it is supplied with a SteriShield Delivery System to maintain sterility. Ideal for hard, non-porous surfaces in controlled environments, Klercide ensures a clean and safe workspace.