Learn More
Ecolab Ecolab Klerwipe Low Residue Peroxide Wipes, Sterile, Non-Corrosive, 200 x 200 mm
Catalog No. NC2607076
Encompass
Ecolab Klerwipe Low Residue Peroxide Pouch Wipes are sterile, 100% polyester wipes for critical cleaning in controlled environments. They are impregnated with Klercide™ Low Residue Peroxide, which cleans without rinsing, ensuring very low residue and minimal contamination risk. Ideal for cleanrooms, isolators, and production vessels, these wipes are packaged in resealable pouches, providing convenience and efficiency for routine or rotational cleaning applications.
Catalog No. NC2607076 Supplier Ecolab Supplier No. 6600625
Ecolab Klerwipe Low Residue Peroxide Pouch Wipes are sterile, 100% polyester wipes for critical cleaning in controlled environments. They are impregnated with Klercide™ Low Residue Peroxide, which cleans without rinsing, ensuring very low residue and minimal contamination risk. Ideal for cleanrooms, isolators, and production vessels, these wipes are packaged in resealable pouches, providing convenience and efficiency for routine or rotational cleaning applications.