Learn More
Aviva Systems Biology Corp KLF1 Antibody - middle region (ARP32810_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32810_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLF1. Alternate names: EKLF, INLU, HBFQTL6ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-014-54777