Learn More
Aviva Systems Biology Corp KLF1 Antibody - middle region (ARP31493_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31493_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLF1. Alternate names: EKLF, INLU, HBFQTL6ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-54244