Learn More
Aviva Systems Biology Corp KLF11 Antibody - N-terminal region (ARP30042_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30042_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLF11. Alternate names: FKLF, FKLF1, MODY7, TIEG2, Tieg3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-54109