Learn More
Aviva Systems Biology Corp Klf11 Antibody - middle region (ARP37579_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37579_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Klf11. It was validated on Western Blot. Klf11,Kruppel-like factor 11,Tieg2; Tieg3; Tieg2b; 9830142A17; D12Ertd427e; Klf11,Krueppel-like factor 11,Krueppel-like factor 11,TIEG-3,TGFB inducible early growth response 2,TGFB inducible early growth response 3,TGFB-inducible early growth response protein 3,TGFB-inducible early growth response protein 2b,transforming growth factor-beta-inducible early growth response protein 3The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 9830142A17, D12Ertd427e, Tieg2, Tieg2b, Tieg3ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-660