Learn More
Aviva Systems Biology Corp KLF14 Antibody - N-terminal region (ARP36137_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36137_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLF14. Alternate names: BTEB5ApplicationWB. Species reactivity: Dog, Human, Pig, Rabbit
Catalog No. 50-014-53035