Learn More
Aviva Systems Biology Corp KLF15 Antibody - N-terminal region (ARP32587_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32587_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF15. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KLF15. Alternate names: KKLFApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54665