Learn More
Aviva Systems Biology Corp KLF16 Antibody - middle region (ARP30079_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30079_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLF16. Alternate names: BTEB4, DRRF, NSLP2ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-54116