Learn More
Aviva Systems Biology Corp KLF17 Antibody - N-terminal region (ARP39987_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39987_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF17. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human KLF17. Alternate names: FLJ40160, ZNF393, Zfp393, RP4-675G8.1ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-53168