Learn More
Aviva Systems Biology Corp KLF2 Antibody - middle region (ARP37859_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37859_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: LklfApplicationWB. Species reactivity: Human, Mouse, Pig, Rat
Catalog No. 50-014-56452