Learn More
Aviva Systems Biology Corp KLF2 Antibody - middle region (ARP32760_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32760_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLF2. Alternate names: LKLFApplicationCHIP, IHC, WB. Species reactivity: Guinea Pig, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-54746