Learn More
Aviva Systems Biology Corp Klf2 Antibody - N-terminal region (ARP36906_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36906_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Klf2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: LklfApplicationWB. Species reactivity: Human, Mouse, Pig, Rat
Catalog No. 50-284-541