Learn More
Aviva Systems Biology Corp Klf3 Antibody - C-terminal region (ARP39264_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39264_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Klf3. It was validated on Western Blot. . Alternate names: Bklf, Tef-2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-794