Learn More
Aviva Systems Biology Corp KLF4 Antibody - N-terminal region (ARP35721_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35721_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLF4. Alternate names: EZF, GKLFApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Sheep
Catalog No. 50-014-55828