Learn More
Aviva Systems Biology Corp Klf4 Antibody - middle region (ARP37076_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37076_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Klf4. It was validated on Western Blot. Klf4,Kruppel-like factor 4 (gut),EZF; Zie; Gklf; Klf4,Krueppel-like factor 4,Krueppel-like factor 4,gut-enriched krueppel-like factor,epithelial zinc finger protein EZF,Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF)The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: EZF, Gklf, ZieApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-284-581