Learn More
Aviva Systems Biology Corp KLF6 Antibody - N-terminal region (ARP35640_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35640_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLF6. Alternate names: BCD1, COPEB, CPBP, DKFZp686N0199, GBF, PAC1, ST12, ZF9, CBA1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55798