Learn More
Aviva Systems Biology Corp KLF9 Antibody - N-terminal region (ARP31369_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31369_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human KLF9. Alternate names: BTEB, BTEB1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54198