Learn More
Bioworld KLH SEPAROPOREAGAROSE) 4B
Catalog No. 50197698
Encompass
KLH-Separopore 4B Affinity matrix with immobilized KLH for the purification of antibodies generated against a peptide-KLH conjugated antigens. It allows removal of antobodies against KLH portion of the antigen; Supplied as a suspension i...
Catalog No. 50-197-698 Supplier Bioworld Supplier No. 201810161
KLH-Separopore 4B Affinity matrix with immobilized KLH for the purification of antibodies generated against a peptide-KLH conjugated antigens. It allows removal of antobodies against KLH portion of the antigen; Supplied as a suspension i...