Learn More
Aviva Systems Biology Corp KLHDC3 Antibody - N-terminal region (ARP52492_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52492_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHDC3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human KLHDC3. Alternate names: PEAS, RP1-20C7.3, dJ20C7.3, hPEASApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-720