Learn More
ABclonal Technology KLHDC3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A13741100UL
Encompass_Preferred
The protein encoded by this gene contains six repeated kelch motifs that are structurally similar to recombination activating gene 2, a protein involved in the activation of the V(D)J recombination. In mouse, this gene is found to be expressed specifically in testis. Its expression in pachytene spermatocytes is localized to cytoplasma and meiotic chromatin, suggesting that this gene may be involved in meiotic recombination.
Catalog No. 50-000-99316