Learn More
Aviva Systems Biology Corp KLHDC5 Antibody - middle region (ARP51678_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51678_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHDC5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLHDC5. Alternate names: KIAA1340, MGC131714, Ctb9, KLHDC5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60584