Learn More
Aviva Systems Biology Corp KLHDC8B Antibody - middle region (ARP48450_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48450_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHDC8B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLHDC8B. Alternate names: FLJ11302, MGC35097ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59829