Learn More
Aviva Systems Biology Corp KLHDC9 Antibody - middle region (ARP51792_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51792_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHDC9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLHDC9. Alternate names: KARCA1, MGC33338, RP11-544M22.9ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse
Catalog No. 50-014-62900