Learn More
Aviva Systems Biology Corp KLHL14 Antibody - N-terminal region (ARP34677_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34677_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL14. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human KLHL14. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52972