Learn More
Aviva Systems Biology Corp KLHL20 Antibody - C-terminal region (ARP55062_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55062_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL20. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human KLHL20. Alternate names: KHLHX, KLEIP, KLHLX, RP3-383J4.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-998