Learn More
Aviva Systems Biology Corp KLHL23 Antibody - middle region (ARP41123_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41123_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL23. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLHL23. Alternate names: DITHP, FLJ37812, MGC22679, MGC2610ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57730